
4-Isopropylphenylhydrazine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:118427-29-5
:
Formula:C₉H₁₅ClN₂
MDL Number:MFCD00013461
:186.69 g/mol
Melting Pt:203 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together