tert-Butyl-3-(iodomethyl)pyrrolidine-1-carboxylate 97%
Catalog # APOSOR965122-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:311.16 g/mol
Storage Temperature:Ambient
CAS Number:479622-36-1
Formula:C₁₀H₁₈INO₂
MDL Number:MFCD08059333