N-Benzyltetrahydro-2H-pyran-4-amine hydrochloride 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:443344-23-8
:
Formula:C₁₂H₁₈ClNO
MDL Number:MFCD11035934
:227.74 g/mol
Storage Temperature:Ambient
Frequently Bought Together