D(-)-Lactic acid
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068311
- Molecular Weight:90.08 g/mol
- Storage Temperature:Freezer
3 Options Available Below
Product Details & Documents
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068311
- Molecular Weight:90.08 g/mol
- Storage Temperature:Freezer



