Order Entry
Netherlands
ContactUsLinkComponent
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride 98%
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride 98%
Catalog # APOSOR960244-1G
Supplier:  Apollo Scientific
CAS Number:  
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride 98%
Catalog # APOSOR960244-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.68 g/mol
Storage Temperature:Ambient
CAS Number:890324-74-0
Formula:C₁₀H₁₂ClN₃O
MDL Number:MFCD13186189