Order Entry
Netherlands
ContactUsLinkComponent
5-(4-Pyridyl)-1H-tetrazole
Irritant
5-(4-Pyridyl)-1H-tetrazole
Catalog # APOSOR9588-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(4-Pyridyl)-1H-tetrazole
Catalog # APOSOR9588-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :147.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:14389-12-9
Formula:C₆H₅N₅
MDL Number:MFCD00068115