2-Phenoxybenzonitrile 98%
Bottle, 1 g
Catalog # APOSOR953387-1G
Supplier: Apollo Scientific
2-Phenoxybenzonitrile 98%
Catalog # APOSOR953387-1G
Supplier: Apollo Scientific
CAS Number: 6476-32-0
Jump to:
Product Details & Documents
- CAS Number:6476-32-0
- Formula:C₁₃H₉NO
- MDL Number:MFCD08271960
- Molecular Weight:195.2167 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



