Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 97%
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 97%
Catalog # APOSOR950952-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 97%
Catalog # APOSOR950952-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.56 g/mol
Storage Temperature:Ambient
CAS Number:84547-85-3
Formula:C₅H₅ClN₂O₂
MDL Number:MFCD01821187