1'-(tert-Butoxycarbonyl)-1,4'-bipiperidine-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:201810-59-5
:
Formula:C₁₆H₂₈N₂O₄
MDL Number:MFCD06408960
:312.41 g/mol
Storage Temperature:Ambient
Frequently Bought Together