Methyl trans-4-hydroxycyclohexanecarboxylate 95%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate
- CAS Number:6125-57-1
- Formula:C₈H₁₄O₃
- MDL Number:MFCD20441963
- Molecular Weight:158.20 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate
- CAS Number:6125-57-1
- Formula:C₈H₁₄O₃
- MDL Number:MFCD20441963
- Molecular Weight:158.20 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



