Order Entry
Netherlands
ContactUsLinkComponent
2-(1H-1,2,4-Triazol-3-yl)pyridine 95%
2-(1H-1,2,4-Triazol-3-yl)pyridine 95%
Catalog # APOSOR930824-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(1H-1,2,4-Triazol-3-yl)pyridine 95%
Catalog # APOSOR930824-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :146.15 g/mol
Storage Temperature:Ambient
CAS Number:23195-62-2
Formula:C₇H₆N₄
MDL Number:MFCD13188589