
Methyl-4-nitro-1H-pyrazole-3-carboxylate 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138786-86-4
:
Formula:C₅H₅N₃O₄
MDL Number:MFCD00505604
:171.11 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together