(S)-(-)-Citronellal 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5949-05-3
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00075588
:154.25 g/mol
Boiling Pt:207 °C
Density:0.851 g/cm³ (25 °C)
Flash Pt:75 °C
Storage Temperature:Ambient
Frequently Bought Together