2-Azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1259489-92-3
:
Formula:C₁₁H₁₉NO₂
MDL Number:MFCD18325169
:197.28 g/mol
Storage Temperature:Ambient
Frequently Bought Together