Order Entry
Netherlands
ContactUsLinkComponent
4-Methoxycinnamonitrile (cis- and trans- mixture) 95%
Irritant
4-Methoxycinnamonitrile (cis- and trans- mixture) 95%
Catalog # APOSOR921138-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methoxycinnamonitrile (cis- and trans- mixture) 95%
Catalog # APOSOR921138-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:127…140 °C/1mmHg
 :159.19 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:28446-68-6
Formula:C₁₀H₉NO
MDL Number:MFCD00001934