Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

Supplier:   Apollo Scientific
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
Supplier:  Apollo Scientific
Certificates
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

5 Options Available Below

Product Details & Documents
  • CAS Number:66185-74-8
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD10566907
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 21,80 €
    21,80 €
    1 * 100 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 24,60 €
    24,60 €
    1 * 250 mg
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 73,80 €
    73,80 €
    1 * 1 g
       73,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 219,00 €
    219,00 €
    1 * 5 g
       219,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 916,00 €
    916,00 €
    1 * 25 g
       916,00 €

    Specifications:
    New Product
    New Product