2-Piperidin-2-yl-1H-benzimidazole dihydrochloride ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51785-23-0
:
Formula:C₁₂H₁₇Cl₂N₃
MDL Number:MFCD08461220
:274.2 g/mol
Storage Temperature:Ambient
Frequently Bought Together