Bestelling invoeren
Netherlands
ContactUsLinkComponent
2-Bromocyclopentanone 95%
2-Bromocyclopentanone 95%
Catalog # APOSOR916795-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromocyclopentanone 95%
Catalog # APOSOR916795-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Verpakking:
    Bottle
  • VPE:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.01 g/mol
Storage Temperature:Kamertemperatuur
CAS Number:21943-50-0
Formula:C₅H₇BrO
MDL Number:MFCD11109944