Order Entry
Netherlands
ContactUsLinkComponent
1-Methyl-5-phenyl-1H-pyrazole-3-carbaldehyde 97%
Irritant
1-Methyl-5-phenyl-1H-pyrazole-3-carbaldehyde 97%
Catalog # APOSOR9158-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Methyl-5-phenyl-1H-pyrazole-3-carbaldehyde 97%
Catalog # APOSOR9158-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:106…108 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:124344-94-1
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD08271935