1-(2-Methoxyethoxy)-4-nitrobenzene 97%
Catalog # APOSOR913195-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:197.1879 g/mol
Storage Temperature:Ambient
CAS Number:22483-40-5
Formula:C₉H₁₁NO₄
MDL Number:MFCD00463497