
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:154970-45-3
:
Formula:C₁₂H₁₈O₂
MDL Number:MFCD18083704
:194.27 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together