
tert-Butyl-2-isopropyl-4-oxopiperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:313950-41-3
:
Formula:C₁₃H₂₃NO₃
MDL Number:MFCD07367731
:241.33 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together