Order Entry
Netherlands
ContactUsLinkComponent
Ethyl-2-methylthiophene-3-carboxylate 98%
Ethyl-2-methylthiophene-3-carboxylate 98%
Catalog # APOSOR904776-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-methylthiophene-3-carboxylate 98%
Catalog # APOSOR904776-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :170.2300 g/mol
Storage Temperature:Ambient
CAS Number:19432-66-7
Formula:C₈H₁₀O₂S
MDL Number:MFCD14698267