Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-2-(methylamino)benzoate
Ethyl-2-(methylamino)benzoate 95%
Ethyl-2-(methylamino)benzoate 95%
APOSOR903561-1G
:
Apollo Scientific
:
Ethyl-2-(methylamino)benzoate 95%
APOSOR903561-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
179.2157 g/mol
:
Ambient
:
35472-56-1
:
C₁₀H₁₃NO₂
:
MFCD11174593