

6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-sulphonyl chloride 95%
Catalog # APOSOR9019-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.65 g/mol
Storage Temperature:Freezer
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:914637-94-8
Formula:C₆H₇ClN₂O₂S
MDL Number:MFCD08235266