

6,7,8,9-Tetrahydro-5H-imidazo-[1,2-a]azepine-3-sulphonyl chloride 95%
Catalog # APOSOR9018-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.71 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
:GHS07
CAS Number:914637-93-7
Formula:C₈H₁₁ClN₂O₂S
MDL Number:MFCD08235265