Order Entry
Netherlands
ContactUsLinkComponent
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 95+%
Irritant
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 95+%
Catalog # APOSOR8641-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 95+%
Catalog # APOSOR8641-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :249.08 g/mol
Melting Pt:175…176 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:852054-42-3
Formula:C₇H₅BrO₃S
MDL Number:MFCD08690306