Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzenecarboxamide
2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzenecarboxamide
Catalogus # APOSOR8499-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-N-(2-hydroxyethyl)-4-(1H-1,2,4-triazol-1-yl)benzenecarboxamide
Catalogus # APOSOR8499-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:140…142 °C
 :266.69 g/mol
Storage Temperature:Ambient
CAS Number:952183-15-2
Formula:C₁₁H₁₁ClN₄O₂
MDL Number:MFCD09607897