[1-(4-Chlorobenzyl)-1H-1,2,3-triazol-4-yl]methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321405-39-4
:
Formula:C₁₀H₁₀ClN₃O
MDL Number:MFCD09607909
:223.66 g/mol
Melting Pt:87…89 °C
Storage Temperature:Ambient
Frequently Bought Together