1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid
Catalog # APOSOR8334-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.7 g/mol
Melting Pt:123…125 °C
Storage Temperature:Ambient
CAS Number:874800-64-3
Formula:C₁₂H₁₃ClN₂O₄
MDL Number:MFCD05664784