Order Entry
Netherlands
ContactUsLinkComponent
Ethyl-4-(3-ethoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Ethyl-4-(3-ethoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Catalog # APOSOR8265-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-4-(3-ethoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Catalog # APOSOR8265-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.33 g/mol
Storage Temperature:Ambient
CAS Number:54278-10-3
Formula:C₁₄H₂₁NO₄
MDL Number:MFCD00010644