
N-Methyl-N-[4-(methylsulphonyl)-2-nitrophenyl]amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:30388-44-4
:
Formula:C₈H₁₀N₂O₄S
MDL Number:MFCD01016949
:230.24 g/mol
Melting Pt:188…190 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together