
3-Bromo-4-chlorobenzoic acid
Catalog # APOSOR7862-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:235.46 g/mol
Melting Pt:218…222 °C
:GHS07
Storage Temperature:Ambient
CAS Number:42860-10-6
Formula:C₇H₄BrClO₂
MDL Number:MFCD00079706