Order Entry
Netherlands
ContactUsLinkComponent
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline 97%
Toxic
Environment
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline 97%
Catalog # APOSOR7811-250MG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Environment
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline 97%
Catalog # APOSOR7811-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :272.31 g/mol
Melting Pt:230 °C (dec.)
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:396129-53-6
Formula:C₁₇H₁₂N₄
MDL Number:MFCD00800758