
4-Butylphenylhydrazine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64287-11-2
:
Formula:C₁₀H₁₇ClN₂
MDL Number:MFCD00800665
Melting Pt:>215 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together