
4-Hydrazinobenzenesulphonamide hydrochloride 99%
Catalog # APOSOR7764-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:223.68 g/mol
Melting Pt:217…219 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:17852-52-7
Formula:C₆H₉N₃O₂S
MDL Number:MFCD00052262