
4-Chloro-N-cyclopropyl-N-(4-piperidinyl)benzene-sulphonamide 95+%
Catalog # APOSOR7412-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:314.84 g/mol
Melting Pt:111…114 °C
:GHS07
Storage Temperature:Ambient
CAS Number:387350-81-4
Formula:C₁₄H₁₉ClN₂O₂S
MDL Number:MFCD01567991