Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-[1,2,4]Triazol-1-ylbutyric acid
4-[1,2,4]Triazol-1-ylbutyric acid
4-[1,2,4]Triazol-1-ylbutyric acid
4-[1,2,4]Triazol-1-ylbutyric acid
APOSOR7105-1G
:
Apollo Scientific
:
4-[1,2,4]Triazol-1-ylbutyric acid
APOSOR7105-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
155.16 g/mol
:
GHS07
:
Ambient
:
158147-52-5
:
C₆H₉N₃O₂
:
MFCD06801317