
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one 97%
Catalog # APOSOR6071-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:226.08 g/mol
Melting Pt:132…133 °C
:GHS07
Storage Temperature:Ambient
CAS Number:20870-90-0
Formula:C₉H₈BrNO
MDL Number:MFCD00464061