

7-Bromo-1-heptene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4117-09-3
:
Formula:C₇H₁₃Br
MDL Number:MFCD01631137
:177.085 g/mol
Boiling Pt:27 °C/1mmHg
Density:1.162 g/cm³ (25 °C)
Flash Pt:58.3 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together