
2-Methanesulphonyl-4,6-dimethoxypyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113583-35-0
:
Formula:C₇H₁₀N₂O₄S
MDL Number:MFCD00672151
:218.23 g/mol
Melting Pt:129…133 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together