
3-(2-Naphthyl)-1H-pyrazole
Catalog # APOSOR5742-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:194.24 g/mol
Melting Pt:158…159 °C
Density:1.302 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:150433-20-8
Formula:C₁₃H₁₀N₂
MDL Number:MFCD01569444