

1-Octyne 96%
Catalogus # APOSOR5513-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:123…126 °C
Melting Pt:–60 °C
Density:0.746 g/cm³ (25 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:211-069-3
UN:3295
ADR:3,II
:110.2 g/mol
:GHS02
:GHS08
CAS Number:629-05-0
Formula:C₈H₁₄
MDL Number:MFCD00009546