Order Entry
Netherlands
ContactUsLinkComponent
Methyl-5-hydroxy-4-methoxy-2-nitrobenzoate 95+%
Irritant
Methyl-5-hydroxy-4-methoxy-2-nitrobenzoate 95+%
Catalog # APOSOR54343-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-5-hydroxy-4-methoxy-2-nitrobenzoate 95+%
Catalog # APOSOR54343-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.18 g/mol
Melting Pt:146…149 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:215659-03-3
Formula:C₉H₉NO₆
MDL Number:MFCD17168870