
3-(N-tert-Butoxycarbonyl-N-methylamino)aniline 97%
Catalog # APOSOR52385-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.28 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:528882-16-8
Formula:C12H18N2O2
MDL Number:MFCD13677178