
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol
Catalog # APOSOR52221-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:113.1200 g/mol
Melting Pt:72…75 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:135242-93-2
Formula:C₄H₇N₃O
MDL Number:MFCD16710296