Gallic acid, anhydrous 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149-91-7
:
Formula:C₇H₆O₅
MDL Number:MFCD00002510
:170.12 g/mol
Melting Pt:251 °C (dec.)
Density:1.694 g/cm³ (25 °C)
EINECS:205-749-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together