
3'-Methoxyacetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:586-37-8
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008736
:150.18 g/mol
Boiling Pt:239…241 °C
Density:1.094 g/cm³ (25 °C)
Flash Pt:110 °C
EINECS:209-573-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together