Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate 97%
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate 97%
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate 97%
APOSOR480800-5G
:
Apollo Scientific
:
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate 97%
APOSOR480800-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
Ambient
:
74420-02-3
:
C₇H₆N₂O·H₂O
:
MFCD08272229