Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 97+%
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 97+%
APOSOR480757-1G
:
Apollo Scientific
:
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 97+%
APOSOR480757-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
163.18 g/mol
:
Ambient
:
916792-12-6
:
C₈H₉N₃O
:
MFCD08060940